Order Entry
Spain
ContactUsLinkComponent
 
Methyl thieno[3,2-b]thiophene-2-carboxylate ≥98.0% (by GC)
 :  TCI
 
undefined
Methyl thieno[3,2-b]thiophene-2-carboxylate ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    96.0 to 100.0 deg-C

 

Frequently Bought Together