2-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5094-12-2
:
Formula:C₁₂H₁₄N₂
MDL Number:MFCD00460783
:186.26 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:170.0 to 174.0 deg-C
Specification Test Results
Frequently Bought Together