5-Methoxyindole-2-carboxylic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4382-54-1
:
Formula:C₁₀H₉NO₃
MDL Number:MFCD00005614
:191.19 g/mol
EINECS:224-487-6
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together