
2-Hydroxypropyl methacrylate (mixture of isomers) ≥97.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:27813-02-1
:923-26-2
Formula:C₇H₁₂O₃
MDL Number:MFCD00051317
Flash Pt:104°C
:Stabilised with 4-methoxyphenol (MEHQ)
:144.17 g/mol
EINECS:248-666-3
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 % (total of isomers)
Specification Test Results
Frequently Bought Together