Order Entry
Spain
ContactUsLinkComponent
Lithium tetrahydroborate ∼4 mol/L in THF
Flammable
Corrosive
Irritant
Health
Lithium tetrahydroborate ∼4 mol/L in THF
Catalog # TCIAL0186-100ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Lithium tetrahydroborate ∼4 mol/L in THF
Catalog # TCIAL0186-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to Brown clear liquid to cloudy liquid
  • Concentration (Gasburet method):
    9.5 to 11.5 w/w%

Specification Test Results

About this item

 :
 :21.78 g/mol
Merck Index:14,05525
Flash Pt:-18°C
EINECS:241-021-7
UN:3399
ADR:4.3,I
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:16949-15-8
Formula:H₄BLi
MDL Number:MFCD00011088
Flash Pt:–18°C