Order Entry
Spain
ContactUsLinkComponent
Lanthionine (DL- and meso- mixture) ≥94.0% (by GC, titration analysis)
Lanthionine (DL- and meso- mixture) ≥94.0% (by GC, titration analysis)
Catalog # TCIAL0010-100MG
Supplier:  TCI
CAS Number:  
Lanthionine (DL- and meso- mixture) ≥94.0% (by GC, titration analysis)
Catalog # TCIAL0010-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (GC):
    min. 94.0 %
  • Purity (Nonaqueous Titration):
    min. 94.0 %

Specification Test Results

About this item

 :
 :208.23 g/mol
Merck Index:14,05365
EINECS:213-076-7
CAS Number:922-55-4
Formula:C₆H₁₂N₂O₄S
MDL Number:MFCD00002615