3,5,7,4'-Tetrahydroxy-3'-methoxyflavone ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:480-19-3
:
Formula:C₁₆H₁₂O₇
MDL Number:MFCD00017310
:316.27 g/mol
EINECS:207-545-5
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Carbon):57.70 to 63.80 %
Specification Test Results
Frequently Bought Together