
Imipramine hydrochloride ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:113-52-0
:
Formula:C₁₉H₂₄N₂·HCl
MDL Number:MFCD00012669
:316.87 g/mol
Merck Index:14,04920
EINECS:204-030-7
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:171.0 to 175.0 deg-C
Specification Test Results
Frequently Bought Together