Order Entry
Spain
ContactUsLinkComponent
 
Flammable
Irritant
Diisopropyl ether ≥98.0% (by GC) stabilised
Catalog # TCIAI0917-500ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Diisopropyl ether ≥98.0% (by GC) stabilised
Catalog # TCIAI0917-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :102.18 g/mol
Merck Index:14,05212
Boiling Pt:68°C
Melting Pt:-60°C
Melting Pt:–60°C
Flash Pt:-28°C
Flash Pt:–28°C
EINECS:203-560-6
UN:1159
ADR:3,II
 :Stabilised with hydroquinone (HQ)
 :GHS02
 :GHS07
CAS Number:108-20-3
Formula:(CH₃)₂CHOCH(CH₃)₂
MDL Number:MFCD00008880