β-Isocupreidine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAI0728-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:-10.0 to -16.0 deg (C=1, EtOH)
Specification Test Results
About this item
:
:310.40 g/mol
Melting Pt:259°C
CAS Number:253430-48-7
Formula:C₁₉H₂₂N₂O₂