Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Reticulocyte stain
Reticulocyte stain, Sigma-Aldrich®
Reticulocyte stain, Sigma-Aldrich®
Reticulocyte stain, Sigma-Aldrich®
Catalog # R4132-120ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Certificates
Expand image
Reticulocyte stain, Sigma-Aldrich®
Catalog # R4132-120ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Certificates
Specifications
Pk:
120 mL
Seq:
2470
Environmentally Preferable:
Specifications
About this item
Product Details
Documentation
:
Storage Temperature:
Ambient
:
1
Product Details
Documentation