
(S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:178306-52-0
:
Formula:C₁₆H₁₆O₄
MDL Number:MFCD11113284
:272.30 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+11.0 to +15.0 deg (C=1.8, EtOH)
Specification Test Results
Frequently Bought Together