Order Entry
Spain
ContactUsLinkComponent
cis-3-Hexenyl-cis-3-hexenoate ≥95.0% (by GC)
cis-3-Hexenyl-cis-3-hexenoate ≥95.0% (by GC)
Catalog # TCIAH0677-5ML
Supplier:  TCI
CAS Number:  
undefined
cis-3-Hexenyl-cis-3-hexenoate ≥95.0% (by GC)
Catalog # TCIAH0677-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :196.29 g/mol
Flash Pt:104°C
EINECS:262-797-3
CAS Number:61444-38-0
Formula:C₁₂H₂₀O₂
MDL Number:MFCD00036652