Order Entry
Spain
ContactUsLinkComponent
Hansa yellow 45.0% (by UV method)
Hansa yellow 45.0% (by UV method)
Catalog # TCIAH0312-25G
Supplier:  TCI
CAS Number:  
Hansa yellow 45.0% (by UV method)
Catalog # TCIAH0312-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Lambda max.:
    410.0 to 415.0 nm (CHCl3)
  • Absorbance:
    min. 0.300 (10mg/L, CHCl3, 410.0 to 415.0 nm)
  • Drying loss:
    max. 1.0 %

Specification Test Results

About this item

 :
 :340.34 g/mol
EINECS:219-730-8
 :GHS07
CAS Number:2512-29-0
Formula:C₁₇H₁₆N₄O₄
MDL Number:MFCD00078234