Fluorene-2-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:922706-40-9
:
Formula:C₁₉H₂₁BO₂
MDL Number:MFCD16294548
:292.18 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:76.0 to 80.0 deg-C
Specification Test Results
Frequently Bought Together