
3-Fluoro-5-hydroxybenzeneboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAF0954-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 114.0 %
Specification Test Results
About this item
:
:155.92 g/mol
:GHS07
CAS Number:871329-82-7
Formula:C₆H₆BFO₃
MDL Number:MFCD07363778