Order Entry
Spain
ContactUsLinkComponent
Fluorescein-5-maleimide ≥97.0% (by HPLC) (contains 2% N,N-Dimethylformamide at maximum)
 
Fluorescein-5-maleimide ≥97.0% (by HPLC) (contains 2% N,N-Dimethylformamide at maximum)
  TCIAF0810-25MG
 :  TCI
 :  
 
Fluorescein-5-maleimide ≥97.0% (by HPLC) (contains 2% N,N-Dimethylformamide at maximum)
  TCIAF0810-25MG
 :  TCI
 :  

 

  • Pk:
    25 mg
  • Seq:
    2315

 

 

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Elemental analysis (Nitrogen):
    3.10 to 3.50 %

 

 

 :
 :427.37 g/mol
 :GHS07
 :Refrigerator
 :75350-46-8
 :C₂₄H₁₃NO₇
 :MFCD00077345