

Fluorescein 6-isothiocyanate (isomer II) ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAF0783-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow to Amber powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
About this item
:
:389.38 g/mol
:GHS07
:GHS08
Storage Temperature:Refrigerator
CAS Number:18861-78-4
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00041838