
Ethyl (11bR)-4-Amino-2,6-bis(3,5-di-tert-butylphenyl)-4,5-dihydro-3H-cyclohepta[1,2-a:7,6-a']dinaphthalene-4-carboxylate ≥97.0% (by HPLC)
Catalog # TCIAE1267-50MG
Supplier: TCI
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
Specification Test Results
About this item
:
:758.10 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1678540-23-2
Formula:C₅₄H₆₃NO₂