
4,4'-(Ethyne-1,2-diyl)diphthalic anhydride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:129808-00-0
:
Formula:C₁₈H₆O₆
MDL Number:MFCD28976167
:318.24 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Morpholine Method):min. 98.0 %
Specification Test Results
Frequently Bought Together