Order Entry
Spain
ContactUsLinkComponent
 
 
 
Ethyl propenyl ether cis- and trans-mixture ≥98.0% (by GC)
 :  TCI
 
 
 
Ethyl propenyl ether cis- and trans-mixture ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomers)

 

Frequently Bought Together