Order Entry
Spain
ContactUsLinkComponent
 
Ethyl-N-(tert-butoxycarbonyl)-D-pyroglutamate ≥95.0% (by GC)
 :  TCI
 
Ethyl-N-(tert-butoxycarbonyl)-D-pyroglutamate ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    54.0 to 58.0 deg-C
  • Specific rotation [a]20/D:
    +44.0 to +47.0 deg (C=1、MeOH)

 

Frequently Bought Together