Order Entry
Spain
ContactUsLinkComponent
Ethyl-6-bromocoumarin-3-carboxylate ≥98.0% (by GC)
Ethyl-6-bromocoumarin-3-carboxylate ≥98.0% (by GC)
Supplier:  TCI
Certificates
Ethyl-6-bromocoumarin-3-carboxylate ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    168.0 to 172.0 deg-C

Specification Test Results

Frequently Bought Together