Order Entry
Spain
ContactUsLinkComponent
Dipentaerythritol Hexakis(3-mercaptopropionate) ≥93.0% (by titrimetric analysis)
Dipentaerythritol Hexakis(3-mercaptopropionate) ≥93.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Dipentaerythritol Hexakis(3-mercaptopropionate) ≥93.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (Iodometric back titration):
    min. 93.0 %

Specification Test Results

Related Information

Frequently Bought Together