Dimethyl 2,2'-Bipyridine-5,5'-dicarboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1762-45-4
:
Formula:C₁₄H₁₂N₂O₄
MDL Number:MFCD30531632
:272.26 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together