Order Entry
Spain
ContactUsLinkComponent
2,6-Dibromo-4,8-bis(n-octyloxy)benzo[1,2-b:4,5-b']dithiophene ≥98.0% (by HPLC)
2,6-Dibromo-4,8-bis(n-octyloxy)benzo[1,2-b:4,5-b']dithiophene ≥98.0% (by HPLC)
Catalog # TCIAD4621-200MG
Supplier:  TCI
CAS Number:  
2,6-Dibromo-4,8-bis(n-octyloxy)benzo[1,2-b:4,5-b']dithiophene ≥98.0% (by HPLC)
Catalog # TCIAD4621-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    61.0 to 65.0 deg-C

Specification Test Results

About this item

 :
 :604.50 g/mol
CAS Number:1294515-75-5
Formula:C₂₆H₃₆Br₂O₂S₂