
4,4'-Dibromo-2,2'-bis(triisopropylsilyl)-5,5'-bithiazole ≥98.0% (by HPLC)
Catalog # TCIAD4605-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):4.10 to 4.60 %
- Melting point:111.0 to 115.0 deg-C
Specification Test Results
About this item
:
:638.71 g/mol
:GHS07
CAS Number:1223559-98-5
Formula:C₂₄H₄₂Br₂N₂S₂Si₂