

Diethylenetriamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-40-0
:
Formula:C₄H₁₃N₃
MDL Number:MFCD00008171
:103.17 g/mol
Boiling Pt:207 °C (1013 hPa)
Melting Pt:–39 °C
Density:0.9523 g/cm³ (20 °C)
Flash Pt:97 °C
UN:2079
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:203-865-4
:GHS06
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-35 °C
- Molar Mass:103.16 g/mol
- Boiling Point:206 - 209 °C (1013 hPa)
- Vapor Pressure:0.5 hPa (20 °C)
- Flash Point:97 °C
- Refractive Index:1.4826 (20 °C, 589 nm)
- Explosion Limit:4.4 - 16.1 %(V)
- Density:0.95 g/cm3 (20 °C)
- pH:12 (100 g/l, H2O, 20 °C)
- Ignition Point:325 °C
- Synonyms:2,2'-Iminobis (ethylamine), Bis(2-aminoethyl)amine, 2,2'-Diaminodiethylamine
- Hill Formula:C4H13N3
Specification Test Results
Related Information
Frequently Bought Together