Order Entry
Spain
ContactUsLinkComponent
1,4-Dihydro-6-methylquinoxaline-2,3-dione ≥98.0% (by HPLC)
1,4-Dihydro-6-methylquinoxaline-2,3-dione ≥98.0% (by HPLC)
Catalog # TCIAD2970-25G
Supplier:  TCI
CAS Number:  
1,4-Dihydro-6-methylquinoxaline-2,3-dione ≥98.0% (by HPLC)
Catalog # TCIAD2970-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 97.0 % (calcd.on anh.substance)
  • Water:
    max. 7.0 %

Specification Test Results

About this item

 :
 :176.18 g/mol
EINECS:228-628-2
CAS Number:6309-61-1
Formula:C₉H₈N₂O₂
MDL Number:MFCD03055725