
1,1-Dimethoxycyclohexane ≥96.0% (by GC)
Catalog # TCIAD1372-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
About this item
:
:144.21 g/mol
Boiling Pt:64°C
Flash Pt:50°C
UN:1993
ADR:3,III
:GHS02
CAS Number:933-40-4
Formula:C₈H₁₆O₂
MDL Number:MFCD00043714