
meso-ɑ,β-Di(4-pyridyl) glycol ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD0936-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:216.24 g/mol
:GHS07
CAS Number:4972-49-0
Formula:C₁₂H₁₂N₂O₂
MDL Number:MFCD00023627