Order Entry
Spain
ContactUsLinkComponent
 
 
2-Chloro-N,N-dimethylacetamide ≥97.0% (by GC)
 :  TCI
 
 
2-Chloro-N,N-dimethylacetamide ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to lump
  • Purity (GC):
    min. 97.0 %

 

Frequently Bought Together