Dipyridin-2-ylcarbonate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1659-31-0
:
Formula:C₁₁H₈N₂O₃
MDL Number:MFCD00191407
:216.1928 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together