tert-Butyl (R)-2-(2,6-Dichlorophenyl)-5-oxo-3-[(R)-1-phenylethyl]imidazolidine-1-carboxylate ≥98.0% (by HPLC)
TCIAB6108-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Optical purity (LC):min. 98.0 ee%
- Melting point:99.0 to 103.0 deg-C
:
:435.35 g/mol
:Refrigerator
:1004755-00-3
:C₂₂H₂₄Cl₂N₂O₃
:MFCD30187600