Fmoc-Dap(Boc)-OH ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:162558-25-0
:
Formula:C₂₃H₂₆N₂O₆
MDL Number:MFCD00235896
:426.47 g/mol
Melting Pt:63°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:-7.0 to -9.0 deg (C=, MeOH) (calcd.on anh.substance)
- Water:max. 3.0 %
Specification Test Results
Frequently Bought Together