Boc-11-Aun-OH ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10436-25-6
:
Formula:C₁₆H₃₁NO₄
MDL Number:MFCD00076902
:301.427 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:66.0 to 70.0 deg-C
Specification Test Results
Frequently Bought Together