Order Entry
Spain
ContactUsLinkComponent
 
2-Methoxy-1-methylethyl acetate (PGMEA)
Catalog # EHERC16527500
Supplier:  EHRENSTORFER
CAS Number:  
2-Methoxy-1-methylethyl acetate (PGMEA)
Catalog # EHERC16527500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :132.16 g/mol
Boiling Pt:145.8 °C (1013 hPa)
Melting Pt:–67 °C
Density:0.968 g/cm³ (20 °C)
Flash Pt:45 °C
EINECS:203-603-9
UN:3272
ADR:3,III
 :GHS02
CAS Number:108-65-6
Formula:C₆H₁₂O₃
MDL Number:MFCD00038500