
2-Bromo-5-nitro-4-picoline ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:23056-47-5
:
Formula:C₆H₅BrN₂O₂
MDL Number:MFCD03095066
:217.02 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:63.0 to 67.0 deg-C
Specification Test Results
Frequently Bought Together