Order Entry
Spain
ContactUsLinkComponent
 
Boc-N-Me-D-Phe-OH ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
Boc-N-Me-D-Phe-OH ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    84.0 to 88.0 deg-C
  • Specific rotation [a]20/D:
    +85.0 to +89.0 deg (C=1, MeOH)

 

Frequently Bought Together