
1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
: TCI
:877399-74-1
:
:C₁₉H₃₂BN₃O₄
:MFCD11112131
:377.2861 g/mol
:GHS07
:Refrigerator
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:114.0 to 118.0 deg-C
Frequently Bought Together