tert-Butyl-3-(hydroxymethyl)piperazine-1-carboxylate ≥98.0% (by GC, titration analysis)
: TCI
:301673-16-5
:
:C₁₀H₂₀N₂O₃
:MFCD04115304
:216.2774 g/mol
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:93.0 to 97.0 deg-C
Frequently Bought Together