Order Entry
Spain
ContactUsLinkComponent
1-tert-Butyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate ≥97.0% (by HPLC, total nitrogen)
Irritant
1-tert-Butyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate ≥97.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
Irritant
1-tert-Butyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate ≥97.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    56.0 to 61.0 deg-C

Specification Test Results

Frequently Bought Together