2-Bromo-4-chlorobenzoic acid ≥96.0% (by GC, titration analysis)
TCIAB4592-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 96.0 %
- Melting point:157.0 to 161.0 deg-C
:
:235.46 g/mol
:GHS07
:936-08-3
:C₇H₄BrClO₂
:MFCD00672930