Order Entry
Spain
ContactUsLinkComponent
5-Bromo-6-hydroxy-1,3-benzodioxole ≥98.0% (by HPLC)
Irritant
5-Bromo-6-hydroxy-1,3-benzodioxole ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
Irritant
5-Bromo-6-hydroxy-1,3-benzodioxole ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    77.0 to 82.0 deg-C

Specification Test Results

Frequently Bought Together