Order Entry
Spain
ContactUsLinkComponent
1-Boc-4-(tosyloxy)piperidine ≥98.0% (by HPLC)
Irritant
1-Boc-4-(tosyloxy)piperidine ≥98.0% (by HPLC)
Catalog # TCIAB4516-1G
Supplier:  TCI
CAS Number:  
Irritant
1-Boc-4-(tosyloxy)piperidine ≥98.0% (by HPLC)
Catalog # TCIAB4516-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 95.0 %
  • Melting point:
    96.0 to 100.0 deg-C

Specification Test Results

About this item

 :
 :355.4500 g/mol
 :GHS07
CAS Number:118811-07-7
Formula:C₁₇H₂₅NO₅S
MDL Number:MFCD06796535