Order Entry
Spain
ContactUsLinkComponent
 
 
4-Bromo-2-(trifluoromethyl)phenol ≥97.0% (by GC)
 :  TCI
 
 
4-Bromo-2-(trifluoromethyl)phenol ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    83.0 to 87.0 deg-C

 

Frequently Bought Together