Order Entry
Spain
ContactUsLinkComponent
4-Bromo-1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-S-indacene ≥95.0% (by GC)
4-Bromo-1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-S-indacene ≥95.0% (by GC)
Supplier:  TCI
Certificates
4-Bromo-1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-S-indacene ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    153.0 to 157.0 deg-C

Specification Test Results

Frequently Bought Together