
Trifluoro(1,1,2,2-tetrafluoro-2-bromoethoxy)ethylene ≥98.0% (by GC) stabilised
Catalog # TCIAB4169-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:276.94 g/mol
:Stabilised with 4-methoxyphenol (MEHQ)
EINECS:288-516-4
:GHS07
Storage Temperature:Refrigerator
CAS Number:85737-06-0
Formula:F₂CCFOCF₂CF₂Br
MDL Number:MFCD02183517