
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate ≥98.0% (by GC, titration analysis)
Catalog # TCIAB4051-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:114.0 to 118.0 deg-C
Specification Test Results
About this item
:
:309.21 g/mol
:GHS07
CAS Number:286961-14-6
Formula:C₁₆H₂₈BNO₄
MDL Number:MFCD03840345