1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:70356-09-1
:
Formula:C₂₀H₂₂O₃
MDL Number:MFCD00210252
Merck Index:14,00888
:310.39 g/mol
EINECS:274-581-6
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:81.0 to 87.0 deg-C
Specification Test Results
Frequently Bought Together